4,7-Dihydroxy-1,10-phenanthroline ≥30% (dye content), Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ100

429031-1G 429031-250MG
429031-1GEA 227 EUR
429031-1G 429031-250MG
4,7-Dihydroxy-1,10-phenanthroline ≥30% (dye content), Sigma-Aldrich®
4,7-Dihydroxy-1,10-phenanthroline

4,7-Dihydroxy-1,10-phenanthroline, Dye content ≥30 % (1,10-Phenanthroline-4,7-diol)

Formule: C₁₂H₈N₂O₂
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00004975
CAS nummer: 3922-40-5
EINECS: 223-493-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff4737047ffbdbe3546147fbfe462cabfa31bd5&vid=8ff4737047ffbdbe3546147fbfe462cabfa31bd5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour): Grey to Brown to Green Conforms to Requirements
Appearance (Form) Powder or Chunks
Infrared spectrum Conforms to Structure
Carbon ≥18.3%
Nitrogen ≥3.9%
Purity (HPLC) Typically >85%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR