Platinum rhodium alloy (94:6 w%), wire for ISA type B standard grade thermocouple, Ø 0.5 mm (0.02 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

42914.BU 42914.BY
42914.BUEA 667 EUR
42914.BU 42914.BY
Platinum rhodium alloy (94:6 w%), wire for ISA type B standard grade thermocouple, Ø 0.5 mm (0.02 in)
Platinum rhodium alloy (94:6 w%)

Molecular Formula: Pt:Rh; 94:6 wt% Physical Form: ≈1.04g/25cm Odour: Odourless MDL No.: MFCD00198186

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR