MULTISURE® HIV Rapid Test, MP Biomedicals

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43030-020
43030-020EA 103 EUR
43030-020
MULTISURE® HIV Rapid Test, MP Biomedicals
Blotkits
The MULTISURE ® HIV rapid test is intended as a rapid diagnostic test, developed to detect antibodies specific to HIV-1 gp120, HIV-1 gp41, HIV-1 p24 (also react with HIV-2) and HIV-2 gp36 antigens in human serum, plasma, finger pricked whole blood or whole blood with anti-coagulants.

  • Diagnostic platforms: Rapid test
  • Diagnostic solutions: Infectious disease
  • Disease screened: Human Immunodeficiency Virus (HIV)
  • Sample type: Plasma, serum, whole blood
  • Analyte/target: HIV 1 and 2 antibodies
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR