Kobalt ≥99.95% (metals basis), rods, Ø 5 mm (0.2 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

43032.DOEA 429 EUR
43032.DO 43032.DJ
Kobalt ≥99.95% (metals basis), rods, Ø 5 mm (0.2 in)
Kobalt

Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈1.75g/cm Odour: Odourless MDL No.: MFCD00010935

Formule: Co
Kookpunt: 2870 °C (1013 hPa)
Smeltpunt: 1495 °C
Dichtheid: 8,9 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010935
CAS nummer: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR