Poly(vinylbenzyl chloride)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

43067.09 43067.18 43067.04
43067.09EA 128 EUR
43067.09 43067.18 43067.04
Poly(vinylbenzyl chloride)
Poly(vinylbenzyl chloride)

CAS No.: 9080-67-5 Molecular Formula: -[CH2CH(C6H4CH2Cl-]n Physical Form: Solid MDL No.: MFCD00135445

Formule: -[CH₂CH(C₆H₄CH₂Cl-]n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00135445
CAS nummer: 9080-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR