Calcium carbonate 99,997 Metals basis, Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Marble

43073.22 43073.14 43073.36 43073.06
43073.22EA 818 EUR
43073.22 43073.14 43073.36 43073.06
Calcium carbonate 99,997 Metals basis, Puratronic®
Calcium carbonate

Purity: 99.997% CAS No.: 471-34-1 Molecular Formula: CaCO3 Formula Weight: 100.09 Physical Form: Powder Density: 2.930 Refractive Index: 1.6583 Appearance: White Odour: Odourless Melting Point: 800° dec. Merck Reference: 14,1657 MDL No.: MFCD00010906

Formule: CaCO₃
MW: 100.09 g/mol
Smeltpunt: 800 °C (ontbinding)
Dichtheid: 2.930
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010906
CAS nummer: 471-34-1
EINECS: 207-439-9
Merck Index: 14,1657

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR