Lanthanum(III) hydroxide ≥99.95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

43123.18 43123.30
43123.18EA 70.7 EUR
43123.18 43123.30
Lanthanum(III) hydroxide ≥99.95%
Lanthanum(III) hydroxide

Purity: 99.95% CAS No.: 14507-19-8 Molecular Formula: La(OH)3 Formula Weight: 189.93 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: dec. MDL No.: MFCD00016160

Formule: H₃LaO₃
MW: 189.93 g/mol
Smeltpunt: Dec.
Dichtheid: 4.28 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016160
CAS nummer: 14507-19-8
EINECS: 238-510-2
UN: 3262
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR