Magnesium oxide ≥99.99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43186.22 43186.14 43186.36
43186.22EA 712 EUR
43186.22 43186.14 43186.36
Magnesium oxide ≥99.99% (metals basis)
Magnesium oxide

Purity: 99.99% CAS No.: 1309-48-4 Molecular Formula: MgO Formula Weight: 40.30 Physical Form: Powder Density: 3.58 Refractive Index: 1.736 Sensitivity: Air Sensitive Odour: Odourless Melting Point: 2852° Boiling Point: 3600° Merck Reference: 14,5677 MDL No.: MFCD00011109

Formule: MgO
Kookpunt: 3600 °C
Smeltpunt: 2852 °C
Dichtheid: 3,58
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 14,5677

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR