Ultrasone vloeistofprocessors, programmeerbaar, Sonicator®
Leverancier: QSONICA LLC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Q700-220 Q500A-220 Q500-220 Q700A-220 Q500AUK-220 Q500UK-220 Q700AUK-220
432-0237EA 6190 EUR
432-0237 432-0234 432-0233 432-0238 QSONQ500AUK-220 QSONQ500UK-220 QSONQ700AUK-220
Ultrasone vloeistofprocessors, programmeerbaar, Sonicator®
Homogenisatoren Ultrasone celscheiders
A range of programmable ultrasonic processors with applications including cell lysis, nanoparticle dispersion, creating emulsions and general homogenisation. Pulses with 20 KHz frequency over a range of 500 to 700 W depending on the model.
  • Q500:
  • Programmable operation by setting time and amplitude
  • Pulse mode prevents heat build up in temperature sensitive samples
  • Digital amplitude/intensity control
  • Real time energy monitoring
The Q700 introduces some additional features designed to ensure reproducibility of results and flexibility of operation.
More Product Information