Sterilised stand-up bags, Whirl-Pak®
Leverancier: NASCO FORT ATKINSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B01364WA B01365WA B01401WA B01450WA B01449WA B01451WA B01542WA
432-0461EA 252 EUR
432-0461 432-0462 432-0463 432-0466 432-0465 432-0467 432-0470
Sterilised stand-up bags, Whirl-Pak®
Zakken Monsterzakken
Works like a bottle without the cost and storage contraints. Self-standing bags make sampling easier. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Gusseted bottom, for self-standing ease
  • Convenient write-on strip for easy labelling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring and closing
  • Opening pull tabs: For easy opening, non slip textured Tyvek
  • Continuous
More Product Information