Sudan orange G, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Solvent Orange 1, 4-(Phenylazo)resorcinol

M-Clarity™ quality level = MQ100

43207-25MG
43207-25MGEA 129 EUR
43207-25MG
Sudan orange G, Supelco®
Sudan orange G
Formule: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Kookpunt: 371
Smeltpunt: 170
MDL Nummer: MFCD00002275
CAS nummer: 2051-85-6
EINECS: 218-131-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cda6d7c0711321b9b98581320b4cd9aa8f2cf0f&vid=c4c09ace74495f130627e0f4943221999fc1f628&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR