Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.075 mm (0.003 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Packaged under argon.

43307.FF 43307.FI
43307.FFEA 130 EUR
43307.FF 43307.FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.075 mm (0.003 in)
Gadolinium

Purity: 99.9% CAS No.: 7440-54-2 Physical Form: ≈0.36g/25x25mm, Packaged under argon Appearance: Gray UN No.: 3178 MDL No.: MFCD00011022

Formule: Gd
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011022
CAS nummer: 7440-54-2
EINECS: 231-162-2
UN: 3178
ADR: 4.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR