Coomassie brilliant blue G-250, Ultrapure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43318.09 43318.04 43318.18
43318.09EA 82.6 EUR
43318.09 43318.04 43318.18
Coomassie brilliant blue G-250, Ultrapure
Coomassie brilliant blue G-250

Protein staining following gel electrophoresis


  • E(m) (595nm, H₂O) >36,300


Brilliant Blue G is used in analytical biochemistry dye and as a P2X7 antagonist. It is also used in protein staining following gel electrophoresis. It is used to prepare the protein reagent for the determination of protein content of the collagenase enzyme isolated from fish waste. It may be employed as stain for the internal limiting membrane (ILM) for the macular hole (MH) and epiretinal membrane (ERM) surgery. It is useful for SDS gels as it readily stains proteins with minimal background color. Protein bands can be visualized during staining. It has been used in the Bradford dye-binding protein assay. A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids. Brilliant Blue G has been used for determining critical micelle concentration of detergents.


Protein staining following gel electrophoresis

Formule: C₄₇H₄₈N₃NaO₇S₂
Dichtheid: >1 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00078482
CAS nummer: 6104-58-1
EINECS: 228-058-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR