Lithium tri-tert-butoxyaluminum hydride 1 M in THF

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Packaged under argon in resealable ChemSeal™ bottles.

43324.AB 43324.AD
43324.ABEA 44.7 EUR
43324.AB 43324.AD
Lithium tri-tert-butoxyaluminum hydride 1 M in THF
Lithium tri-tert-butoxyaluminum hydride

Packaged under Argon in resealable ChemSeal™ bottles


CAS No.: 17476-04-9 Molecular Formula: C12H28AlLiO3 Formula Weight: 254.28 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.904 Sensitivity: Moisture Sensitive Appearance: Colourless UN No.: UN3399 MDL No.: MFCD00011532

Formule: C₁₂H₂₈AlLiO₃
Dichtheid: 0,904
Vlampunt: –17 °C (1 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011532
CAS nummer: 17476-04-9
UN: 3399
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR