Magnesium ≥99.9% (metals basis), rods, Length: 25 mm (1 in), Ø 7.9 mm (0.31 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43355.KF 43355.KM 43355.KJ
43355.KFEA 39.8 EUR
43355.KF 43355.KM 43355.KJ
Magnesium ≥99.9% (metals basis), rods, Length: 25 mm (1 in), Ø 7.9 mm (0.31 in)
Magnesium

Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈2.1g/piece Odour: Odourless MDL No.: MFCD00085308

Formule: Mg
Kookpunt: 1107 °C (1013 hPa)
Smeltpunt: 651 °C
Dichtheid: 1,74 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00085308
CAS nummer: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR