Iridium(III) chloride, anhydrous ≥99.99% (metals basis) Ir ≥ 63.9%, Premion®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

43380.03 43380.06
43380.03EA 522 EUR
43380.03 43380.06
Iridium(III) chloride, anhydrous ≥99.99% (metals basis) Ir ≥ 63.9%, Premion®
Iridium(III) chloride

Purity: 99.99% CAS No.: 10025-83-9 Molecular Formula: IrCl3 Formula Weight: 298.56 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Green to brown Melting Point: 763° dec. Merck Reference: 14,5088 UN No.: 3260 MDL No.: MFCD00011063

Formule: Cl₃Ir
MW: 298.58 g/mol
Smeltpunt: 763 °C (ontbinding)
Dichtheid: 5.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011063
CAS nummer: 10025-83-9
EINECS: 233-044-6
UN: 3260
ADR: 8 II
Merck Index: 14,5088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR