Iridium(IV) oxide ≥99.99% (metals basis) Ir ≥ 84.5%, Premion®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

43396.06 43396.03
43396.06EA 3016 EUR
43396.06 43396.03
Iridium(IV) oxide ≥99.99% (metals basis) Ir ≥ 84.5%, Premion®
Iridium(IV) oxide

Purity: 99.99% CAS No.: 12030-49-8 Molecular Formula: IrO2 Formula Weight: 224.20 Physical Form: Powder Density: 11.7 Appearance: Black Odour: Odourless UN No.: 1479 MDL No.: MFCD00011065

Formule: IrO₂
Kookpunt: –27 °C (1013 hPa)
Smeltpunt: –102 °C
Dichtheid: 11,7
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011065
CAS nummer: 12030-49-8
EINECS: 234-743-9
UN: 1479
ADR: 5.1,II

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR