Potassium chloride 3 M in aqueous solution

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43398.K2 43398.AK
43398.K2EA 95.7 EUR
43398.K2 43398.AK
Potassium chloride 3 M in aqueous solution
Potassium chloride

CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00011360


For refilling calomel electrodes

Formule: KCl
Dichtheid: 1,13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011360
CAS nummer: 7447-40-7
EINECS: 231-211-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR