Sodium iodide, ultra dry ≥99.98% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Ampuled under argon

43447.22 43447.14 43447.36
43447.22EA 316 EUR
43447.22 43447.14 43447.36
Sodium iodide, ultra dry ≥99.98% (metals basis)
Sodium iodide

Purity: 99.98% CAS No.: 7681-82-5 Molecular Formula: NaI Formula Weight: 149.89 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.67 Refractive Index: 1.7745 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 651° Boiling Point: 1300-1304° Merck Reference: 14,8631 MDL No.: MFCD00003532

Formule: NaI
Kookpunt: 1300…1304 °C
Smeltpunt: 651 °C
Dichtheid: 3,67
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003532
CAS nummer: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 14,8631

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR