Glycine ≥98.5% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43497.22 43497.A5 43497.36
43497.22EA 32 EUR
43497.22 43497.A5 43497.36
Glycine ≥98.5% ACS
Glycine

Purity: 98.5+% CAS No.: 56-40-6 Molecular Formula: NH2CH2CO2H Formula Weight: 75.07 Physical Form: Crystalline Density: 1.595 Appearance: White to off-white Odour: Odourless pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131

Formule: NH₂CH₂COOH
Kookpunt: 181 °C (1013 hPa)
Dichtheid: 1,595
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008131
CAS nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR