Calciumsulfide ≥99% (metals basis excluding Mg) (max. 1% Mg)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

43646.14
43646.14EA 246 EUR
43646.14
Calciumsulfide ≥99% (metals basis excluding Mg) (max. 1% Mg)
Calciumsulfide

Purity: 99% CAS No.: 20548-54-3 Molecular Formula: CaS Formula Weight: 72.14 Physical Form: -100 Mesh Powder Density: 2.5 Appearance: White Odour: Stench Melting Point: dec. Merck Reference: 14,1707 UN No.: 3134 MDL No.: MFCD00015990

Formule: CaS
Smeltpunt: Dec.
Dichtheid: 2,5
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00015990
CAS nummer: 20548-54-3
EINECS: 243-873-5
UN: 3134
ADR: 4.3,III
Merck Index: 14,1707

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR