Leuco malachite green

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43758.09 43758.04
43758.09EA 109 EUR
43758.09 43758.04
Leuco malachite green
Leuco malachite green

CAS No.: 129-73-7 Molecular Formula: C23H26N2 Formula Weight: 330.47 Physical Form: Powder Sensitivity: Air Sensitive Melting Point: 100-102° Beilstein Reference: 2140506 MDL No.: MFCD00008315

Formule: C₂₃H₂₆N₂
Smeltpunt: 100…102 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008315
CAS nummer: 129-73-7
EINECS: 204-961-9
Merck Index: 14,5699

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR