Aluminium oxide, intermediate surface area catalyst support low SiO₂

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43857.A1 43857.A7
43857.A1EA 208 EUR
43857.A1 43857.A7
Aluminium oxide, intermediate surface area catalyst support low SiO₂
Aluminium oxide

Physical Form: 1/8" Pellets MDL No.: MFCD00003424

Formule: Al₂O₃
Kookpunt: 2980 °C (1013 hPa)
Smeltpunt: 2045 °C
Dichtheid: 3,94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Bereik: 01-2119529248-35
Merck Index: 13,00355

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR