Magnesium tetraborate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43892.14 43892.22
43892.14EA 137 EUR
43892.14 43892.22
Magnesium tetraborate ≥99%
Magnesium tetraborate

Purity: 99% CAS No.: 12007-62-4 Molecular Formula: MgB4O7 Formula Weight: 179.54 Physical Form: Powder Odour: Odourless MDL No.: MFCD02688001

Formule: MgB₄O₇
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02688001
CAS nummer: 12007-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR