Bismuth(III) telluride ≥99.98% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Dibismuth tritelluride

44077.14 44077.22
44077.14EA 306 EUR
44077.14 44077.22
Bismuth(III) telluride ≥99.98% (metals basis)
Bismuth(III) telluride

Purity: 99.98% CAS No.: 1304-82-1 Molecular Formula: Bi2Te3 Formula Weight: 800.76 Physical Form: Powder Appearance: Grey Odour: Odourless Melting Point: 573° UN No.: 3284 MDL No.: MFCD00014201

Formule: Bi₂Te₃
Smeltpunt: 573 °C
Dichtheid: 7,69 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014201
CAS nummer: 1304-82-1
EINECS: 215-135-2
UN: 3284
ADR: 6.1,III
Merck Index: 12,01331

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR