Lithium-tert-butanolate ≥99.9% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

44133.04 44133.09 44133.18
44133.04EA 53.8 EUR
44133.04 44133.09 44133.18
Lithium-tert-butanolate ≥99.9% (metals basis)
Lithium-tert-butanolate

Purity: 99.9% CAS No.: 1907-33-1 Molecular Formula: C4H9LiO Formula Weight: 80.06 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White to cream Odour: Odourless Melting Point: ca 150° dec. Beilstein Reference: 3620018 UN No.: 3206 MDL No.: MFCD00050479

Formule: C₄H₉LiO
MW: 80.06 g/mol
Smeltpunt: ∼ 150 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00050479
CAS nummer: 1907-33-1
EINECS: 217-611-5
UN: 3206
ADR: 4.2 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR