HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% for biochemistry

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

441485HEA 140 EUR
441485H 441487M 44148BC
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) ≥99.5% for biochemistry
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid)
Formule: C₈H₁₈N₂O₄S
Smeltpunt: 210…215 °C
Dichtheid: 1,448 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006158
CAS nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Appearance White powder
Appearance of solution (1 mol/l) Clear colourless liquid
IR Spectrum Passes test
pH (1 %) 4.7 - 5.3
Ignition residue Max. 0.3 %
Water Max. 0.5 %
Cl (Chloride) Max. 300 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 100 ppm
Al (Aluminium) Max. 5 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 0.1 %
Pb (Lead) Max. 5 ppm
Absorbance (260 nm) (1 mol/l) Max. 0.10
Absorbance (280 nm) (1 mol/l) Max. 0.10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR