Heat-on blocks
Leverancier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
505-80067-00 505-80069-00 505-80071-00 505-80073-00 505-80075-00 505-81200-00 505-80067-01 505-80082-00 505-80080-00
442-0636EA 466 EUR
442-0636 442-0637 442-0638 442-0639 442-0640 442-0658 HEID505-80067-01 HEID505-80082-00 HEID505-80080-00
Heat-on blocks
Blokverwarmers Blokken
Safe, fast and efficient way of heating and stirring round bottom flasks from 10 ml to 5 litres, suitable for use with all makes of hotplates with diameter up to 145 mm. Heat-On blocks replace oil baths and heating mantles, manufactured from solid aluminium they provide excellent heat transfer, prevent hotspots and avoid spills making chemistry safer. Unique well design prevents flasks cracking whilst maximising surface area contact.
  • Save money by removing the need for messy oil and the costly disposal of used oil, and utilise existing laboratory glassware
More Product Information