Calcium iodide, ultra dry ≥99.999% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Ampuled under argon

44254.14 44254.22
44254.14EA 209 EUR
44254.14 44254.22
Calcium iodide, ultra dry ≥99.999% (metals basis)
Calcium iodide

Purity: 99.999% CAS No.: 10102-68-8 Molecular Formula: CaI2 Formula Weight: 293.89 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.956 Appearance: White to pale yellow Odour: Odourless Melting Point: 784° Boiling Point: 1100° Merck Reference: 14,1677 MDL No.: MFCD00010910

Formule: CaI₂
Kookpunt: 1100 °C
Smeltpunt: 784 °C
Dichtheid: 3,956
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010910
CAS nummer: 10102-68-8
EINECS: 233-276-8
Merck Index: 14,1677

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR