n-Pentaan, dry_extra over Molecular Sieve 99+%, AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Pentane

442990010 442991000
442990010.EA 121 EUR
442990010. 442991000.
n-Pentaan, dry_extra over Molecular Sieve 99+%, AcroSeal®
n-Pentaan
Formule: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Kookpunt: 35…36 °C (1013 hPa)
Smeltpunt: –130 °C
Dichtheid: 0.626 g/cm³
Vlampunt: –49 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009498
CAS nummer: 109-66-0
UN: 1265
ADR: 3 II
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR