Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Aluminum

44326.09 44326.18
44326.09EA 133 EUR
44326.09 44326.18
Aluminium ≥99.9999% (metals basis), slug, Puratronic®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Aluminium

Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈0.07g/slug Odour: Odourless MDL No.: MFCD00134029

Formule: Al
MW: 26.98 g/mol
Kookpunt: 2327
Smeltpunt: 660
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR