Cobalt(II) acetate tetrahydrate 98.0-102.0% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Cobaltous acetate tetrahydrate, Acetic acid cobalt (II) salt tetrahydrate, Cobalt(Ⅱ) acetate tetrahydrate, Cobalt diacetate tetrahydrate

44344.14 44344.22 44344.36
44344.14EA 40.8 EUR
44344.14 44344.22 44344.36
Cobalt(II) acetate tetrahydrate 98.0-102.0% ACS
Cobalt(II) acetate tetrahydrate

Purity: 98.0-102.0% CAS No.: 6147-53-1 Molecular Formula: C4H6CoO4.4H2O Formula Weight: 249.08 (177.02anhy) Physical Form: Crystalline Density: 1.71 Refractive Index: 1.542 Sensitivity: Hygroscopic Appearance: Red Odour: Odourless Melting Point: ca 140° -4H2O Beilstein Reference: 3731397 Merck Reference: 14,2433 UN No.: 3077 MDL No.: MFCD00149569

Formule: (H₃CCOO)₂Co·4H₂O
MW: 249.08 g/mol
Smeltpunt: ∼140 °C (-4H₂O)
Dichtheid: 1.71
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149569
CAS nummer: 6147-53-1
EINECS: 200-755-8
UN: 3077
ADR: 9 III
Merck Index: 14,2433

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR