Trityl chloride resin 0,5 - 2,5 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

44397.03 44397.06 44397.14
44397.03EA 51.5 EUR
44397.03 44397.06 44397.14
Trityl chloride resin 0,5 - 2,5 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Trityl chloride resin

Physical Form: Beads Sensitivity: Hygroscopic Appearance: White to off-white UN No.: UN3261 MDL No.: MFCD00284810

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00284810
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR