Cesium acetate ≥99.998% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

44434.14 44434.22
44434.14EA 111 EUR
44434.14 44434.22
Cesium acetate ≥99.998% (metals basis)
Cesium acetate

Purity: 99.998% CAS No.: 3396-11-0 Molecular Formula: CsOOCCH3 Formula Weight: 191.95 Physical Form: Lump/Powder Odour: Weak MDL No.: MFCD00013056

Formule: H₃CCOOCs
Smeltpunt: 194 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013056
CAS nummer: 3396-11-0
EINECS: 222-248-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR