Manganese(II) chloride tetrahydrate ≥99.99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

44442.06 44442.14 44442.22
44442.06EA 41.8 EUR
44442.06 44442.14 44442.22
Manganese(II) chloride tetrahydrate ≥99.99% (metals basis)
Manganese(II) chloride tetrahydrate

Purity: 99.99% CAS No.: 13446-34-9 Molecular Formula: MnCl2.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Density: 2.010 Melting Point: 58° Merck Reference: 14,5728 MDL No.: MFCD00149792

Formule: MnCl₂·4H₂O
Kookpunt: 1190 °C (1013 hPa)
Smeltpunt: 58 °C
Dichtheid: 2,010
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149792
CAS nummer: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 14,5728

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR