Praseodymium(III,IV) oxide ≥99% (REO, rare earth oxide basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

44594.30 44594.A1
44594.30EA 131 EUR
44594.30 44594.A1
Praseodymium(III,IV) oxide ≥99% (REO, rare earth oxide basis)
Praseodymium(III,IV) oxide

Purity: 99% CAS No.: 12037-29-5 Molecular Formula: Pr6O11 Formula Weight: 1021.43 Physical Form: Powder Melting Point: 2500° MDL No.: MFCD00011178

Formule: O₁₁Pr₆
Smeltpunt: 2500 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011178
CAS nummer: 12037-29-5
EINECS: 234-857-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR