J.T.Baker® BAKER-flex®, Precoated flexible TLC sheet
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4463-02EA 389.58 EUR
4463-02 4463-04
J.T.Baker® BAKER-flex®, Precoated flexible TLC sheet
TLC-platen
Ready-to-use in thin layer chromatography applications, Baker-flex® precoated flexible plastic sheet have a hard, abrasion-resistant coating and solvent-resistant substrate for rapid, clean separations.
The sheets can be written on and cut to size. They are available with or without fluorescent indicator.