Platinum iridium alloy (80:20 w%), wire, hard, Ø 1.0 mm (0.04 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

44732.BQ 44732.BS 44732.BW
44732.BQEA 600 EUR
44732.BQ 44732.BS 44732.BW
Platinum iridium alloy (80:20 w%), wire, hard, Ø 1.0 mm (0.04 in)
Platinum iridium alloy (80:20 w%)

Molecular Formula: Pt:Ir; 80:20 wt% Physical Form: ≈0.85g/5cm Odour: Odourless MDL No.: MFCD02091683

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR