Tetrahydrofuraan, extra dry 99.85% stabilised, AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Butylene oxide, THF, Diethylene oxide

448361000
448361000.EA 59.9 EUR
448361000.
Tetrahydrofuraan, extra dry 99.85% stabilised, AcroSeal®
Tetrahydrofuraan

package of 4x25ML bottles

Formule: C₄H₈O
Kookpunt: 66 °C (760 mmHg)
Dichtheid: 0,88 g/cm³
Vlampunt: –21 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005356
CAS nummer: 109-99-9
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.005 % (Coulometric)
GC ≥99.80 %
Stabilizer ≥200 ppm BHT
Residue after evaporation ≤0.001 %
Filt.over 0.2 micron PFTE Filtered
Acidity (CH3COOH) ≤0.0005 meq/g
Peroxides ≤0.0025 % (H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR