Salpeterzuur 69%, ARISTAR® for trace analysis

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Nitric acid solution

450041MEA 95.2 EUR
450041M 450042N 450043X
Salpeterzuur 69%, ARISTAR® for trace analysis
Salpeterzuur
Formule: HNO₃
Kookpunt: 120,5 °C (1013 hPa)
Smeltpunt: –42 °C
Dichtheid: 1,4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011349
CAS nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1),II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 68.5 - 69.5 %
Cl (Chloride) Max. 0.05 ppm
PO4 (Phosphate) Max. 0.1 ppm
Si (as silicate) Max. 0.1 ppm
SO4 (Sulphate) Max. 0.5 ppm
As + Sb (as As) Max. 0.005 ppm
Ag (Silver) Max. 0.005 ppm
Al (Aluminium) Max. 0.020 ppm
Au (Gold) Max. 0.005 ppm
B (Boron) Max. 0.005 ppm
Ba (Barium) Max. 0.005 ppm
Be (Beryllium) Max. 0.005 ppm
Bi (Bismuth) Max. 0.005 ppm
Ca (Calcium) Max. 0.05 ppm
Cd (Cadmium) Max. 0.005 ppm
Co (Cobalt) Max. 0.005 ppm
Cr (Chromium) Max. 0.010 ppm
Cu (Copper) Max. 0.005 ppm
Fe (Iron) Max. 0.020 ppm
Ga (Gallium) Max. 0.020 ppm
Ge (Germanium) Max. 0.010 ppm
Hg (Mercury) Max. 0.001 ppm
In (Indium) Max. 0.010 ppm
K (Potassium) Max. 0.010 ppm
Li (Lithium) Max. 0.005 ppm
Mg (Magnesium) Max. 0.010 ppm
Mn (Manganese) Max. 0.005 ppm
Mo (Molybdenum) Max. 0.005 ppm
Na (Sodium) Max. 0.050 ppm
Ni (Nickel) Max. 0.005 ppm
Pb (Lead) Max. 0.005 ppm
Pt (Platinum) Max. 0.005 ppm
Sn (Tin) Max. 0.020 ppm
Sr (Strontium) Max. 0.005 ppm
Ti (Titanium) Max. 0.005 ppm
Tl (Thallium) Max. 0.005 ppm
V (Vanadium) Max. 0.005 ppm
Zn (Zinc) Max. 0.010 ppm
Zr (Zirconium) Max. 0.005 ppm
Ignition residue (SO4) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR