Polydimethylsiloxane emulsion

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45088.A4 45088.36 45088.22
45088.A4EA 199 EUR
45088.A4 45088.36 45088.22
Polydimethylsiloxane emulsion
Polydimethylsiloxane
Formule: (CH₃)₃SiO(Si(CH₃)₂O)n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00084411
CAS nummer: 63148-62-9

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR