Scandium 15.9 mm (0.63 in) Ø

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45118.KF
45118.KFEA 298 EUR
45118.KF
Scandium 15.9 mm (0.63 in) Ø
Scandium

CAS No.: 7440-20-2 Physical Form: ≈5g/piece Odour: Odourless MDL No.: MFCD00016323

Formule: Sc
Kookpunt: 2730 °C (1013 hPa)
Smeltpunt: 1541 °C
Dichtheid: 2,989 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016323
CAS nummer: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR