Palladium 5% on activated carbon paste (50% water) 5R58

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45132.22 45132.14 45132.06
45132.22EA 1557 EUR
45132.22 45132.14 45132.06
Palladium 5% on activated carbon paste (50% water) 5R58
Palladium

CAS No.: 7440-05-3 Appearance: Black Odour: Odourless MDL No.: MFCD03457879

Formule: Pd
MW: 106.420 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03457879
CAS nummer: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR