Nickel copper alloy (67:33 w%), alloy 400, foil, Thickness: 1.6 mm (0.062 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

45231.GTEA 196 EUR
45231.GT
Nickel copper alloy (67:33 w%), alloy 400, foil, Thickness: 1.6 mm (0.062 in)
Nickel copper alloy (67:33 w%), alloy 400

Molecular Formula: Ni:Cu; 67:33 wt% MDL No.: MFCD00801098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR