Indium standard solution, 10 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45256.AEEA 86.1 EUR
45256.AE
Indium standard solution, 10 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Standaarden Kationische standaarden Indium standaarden

Molecular Formula: In in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048


Certificaten: Traceable to NIST

Order Now

Specificaties

Environmentally Preferable
MDL No. MFCD00134048

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR