Titanium Standard Solutions for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35759.ADEA 86.2 EUR
35759.AD 35759.AE 35759.AP 35768.AE 38720.AD 38720.AE 38725.AP 35768.AD 35768.AP 38720.AP 38725.AD 38725.AE 45267.AE
Titanium Standard Solutions for ICP, Specpure®
Standaarden Kationische standaarden Titanium standaarden

UN No.: 3264
MDL No.: MFCD00011264


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR