Verwarmingsplaten
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HP1-1 HP1-2 HP1-3
453-0004EA 1880 EUR
453-0004 453-0006 453-0008
Verwarmingsplaten
Verwarmers Verwarmingsplaten
Stainless steel casing, stove enamelled finish and cast aluminium plate. Units accept retort rods.
  • Temperature controlled by hydraulic thermostat
  • With illuminated on/off switch and heater “ON” indicator
  • Fitted with non-slip rubber feet for safety
Each hotplate accepts an optional sand bath attachment with aluminium upstand rim which fits over edge of hotplate. The attachment can also act as a safety rim to prevent dishes falling off hotplate.