Heating tapes, IT-S20

Leverancier: ISOPAD
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

114346-000 054874-000 261076-000 704368-000 153620-000 443830-000
454-3011EA 192.6 EUR
454-3011 ISOP054874-000 ISOP261076-000 ISOP704368-000 ISOP153620-000 ISOP443830-000
Heating tapes, IT-S20
Verwarmers
Isopad IT-S20 is a glass silk, preterminated, flexible heating tape with a copper-tinned outer braiding serving as a protective earth braid. The temperature depends on the power and application. This enables maximum power to be installed even in the small surfaces. Provided with class I electrical protection.

  • Maximum operating temperature: 450 °C
  • Width×Thickness: 12×7 mm (±10%)
  • Maximum power per meter 220 W/m

It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR