VWR® ARISTAR®, Cobalt Standard Solution, 10000 mg/L Co in dil. Nitric Acid (from Co), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
455274BEA 896 EUR
455274B 455272W
VWR® ARISTAR®, Cobalt Standard Solution, 10000 mg/L Co in dil. Nitric Acid (from Co), Standard for ICP
Standaarden Kationische standaarden Kobalt standaarden
Co in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.