2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

45531.03 45531.06
45531.03EA 115 EUR
45531.03 45531.06
2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98%
2,3,4,6-Tetra-O-benzyl-D-galactopyranose

Purity: 98% CAS No.: 53081-25-7 Molecular Formula: C34H36O6 Formula Weight: 540.65 Storage Temperature: -30°C to -10°C Physical Form: Crystalline Appearance: White MDL No.: MFCD00132927

Formule: C₃₄H₃₆O₆
MDL Nummer: MFCD00132927
CAS nummer: 53081-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR